Learn More
Fluotics PerkinElmer Janus Varispan compatible P200, sterile
Supplier:  Fluotics PKE200CB.ST
Encompass
PerkinElmer Janus Varispan compatible P200, sterile, Working Volume: 200uL, Tip Color: Black, Tip Box, Varispan, Compare To: 6001250, 50 racks/case with 96 tips per rack
Catalog No. 50-004-35231