Learn More
Fluotics PerkinElmer Janus Varispan compatible P200, sterile
Catalog No. 5000435252
Encompass
PerkinElmer Janus Varispan compatible P200, sterile, Working Volume: 200uL, Tip Color: Clear, Tip Box, Varispan, 50 racks/case with 96 tips per rack
Catalog No. 50-004-35252 Supplier Fluotics Supplier No. PKE200.ST
PerkinElmer Janus Varispan compatible P200, sterile, Working Volume: 200uL, Tip Color: Clear, Tip Box, Varispan, 50 racks/case with 96 tips per rack