Learn More
Fluotics PerkinElmer Janus compatible P235, filter/sterile
Catalog No. 5000435153
Encompass
PerkinElmer Janus compatible P235, filter/sterile, Working Volume: 180uL, Tip Color: Clear, Tip Box, MDT, Compare To: 6001289, 6073643, 50 racks/case with 96 tips per rack
Catalog No. 50-004-35153 Supplier Fluotics Supplier No. PKE235.F
PerkinElmer Janus compatible P235, filter/sterile, Working Volume: 180uL, Tip Color: Clear, Tip Box, MDT, Compare To: 6001289, 6073643, 50 racks/case with 96 tips per rack