Learn More
Fluotics PerkinElmer Janus Varispan compatible P50, filter/sterile
Supplier:  Fluotics PKEH50.F
Encompass
PerkinElmer Janus Varispan compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Clear, Hanging Rack, Varispan, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35144