Learn More
Fluotics PerkinElmer Janus Varispan compatible P50, filter/sterile
Catalog No. 5000435142
Encompass
PerkinElmer Janus Varispan compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Black, Hanging Rack, Varispan, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35142 Supplier Fluotics Supplier No. PKEH50CB.F
PerkinElmer Janus Varispan compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Black, Hanging Rack, Varispan, 24 racks/case with 96 tips per rack