Learn More
Fluotics Qiagen QIAgility Compatible P200, filter/sterile
Catalog No. 5000435280
Encompass
Qiagen QIAgility Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Black, Hanging Rack, Qiagility standard, Compare To: 990522, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35280 Supplier Fluotics Supplier No. QQ200CB.F
Qiagen QIAgility Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Black, Hanging Rack, Qiagility standard, Compare To: 990522, 24 racks/case with 96 tips per rack