Learn More
Perkin Elmer US LLC PerkinElmer O-Ring, Viton, Glass Liner, 590/690 Injector, Pkg. 10
Catalog No. NC3974901
Encompass
This Viton O-ring is designed for glass liners used in 590/690 cap injectors. It is recommended for mass spectrometry applications and can withstand a maximum injector temperature of 300°C.
  • Made from Viton material
  • For glass liners in 590/690 cap injectors
  • Maximum injector temperature of 300°C
  • Recommended for use with mass spectrometry
  • Comes in a package of 10
Catalog No. NC3974901 Supplier Perkin Elmer US LLC Supplier No. 09200714
This Viton O-ring is designed for glass liners used in 590/690 cap injectors. It is recommended for mass spectrometry applications and can withstand a maximum injector temperature of 300°C.
  • Made from Viton material
  • For glass liners in 590/690 cap injectors
  • Maximum injector temperature of 300°C
  • Recommended for use with mass spectrometry
  • Comes in a package of 10