Learn More
Perkin Elmer US LLC PerkinElmer Pure I, 5% HNO3, 125 mL
Catalog No. 502104221
Encompass_Preferred
Concentration of Element includes: Concentration Element 400 µg/mL Tl 200 µg/mL Bi, ln, Pb 150 µg/mL Ga 100 µg/mL Al 50 µg/mL Ag, Ni 25 µg/mL Cr 20 µg/mL Cd, Co, Cu, Zn 15 µg/mL B, Fe 5 µg/mL Ba, Mn 1µg/mL Be, Sr
Catalog No. 50-210-4221 Supplier Perkin Elmer US LLC Supplier No. N9303940
Add to Cart
Add to Cart
Concentration of Element includes: Concentration Element 400 µg/mL Tl 200 µg/mL Bi, ln, Pb 150 µg/mL Ga 100 µg/mL Al 50 µg/mL Ag, Ni 25 µg/mL Cr 20 µg/mL Cd, Co, Cu, Zn 15 µg/mL B, Fe 5 µg/mL Ba, Mn 1µg/mL Be, Sr