Learn More
Perkin Elmer US LLC PerkinElmer Pure VIII, 5% HNO3, 125 mL
Catalog No. 502104512
Encompass_Preferred
Details Concentration Element 100 µg/mL Al, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Se, Sr, Te, Tl, Zn
Catalog No. 50-210-4512 Supplier Perkin Elmer US LLC Supplier No. N9303942
Add to Cart
Add to Cart
Details Concentration Element 100 µg/mL Al, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Se, Sr, Te, Tl, Zn