Learn More
Aviva Systems Biology Corp PERLD1 Antibody - N-terminal region (ARP50188_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50188_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PERLD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PERLD1. Alternate names: AGLA546, CAB2, MGC9753, PP1498, PER1, PERLD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60278