Learn More
Biotium Permeabilization and Blocking Buffer (5X)
Catalog No. 501964397
Encompass_Preferred
Concentrated buffer stock solution (5X) for one step permeabilization and blocking for immunofluorescence staining. Note: this buffer contains goat serum and therefore is not compatible with anti-goat and anti-sheep secondary antibodies. We recommend Permeabilization Buffer for use with anti-goat and anti-sheep secondary antibodies.
Catalog No. 50-196-4397 Supplier Biotium Supplier No. 22017
Concentrated buffer stock solution (5X) for one step permeabilization and blocking for immunofluorescence staining. Note: this buffer contains goat serum and therefore is not compatible with anti-goat and anti-sheep secondary antibodies. We recommend Permeabilization Buffer for use with anti-goat and anti-sheep secondary antibodies.