Learn More
GA International Permanent Cryo-LazrTAG™- Cryogenic Labels for Laser Printers (US Letter Size) PATENTED
Supplier:  GA International CL3T1BL
Encompass_Preferred
These thermoplastic waterproof labels are specifically designed to permanently adhere to vials, microtubes, microplates, glass tubes, freezer boxes, polycarbonate cryo boxes, and other containers that will undergo long-term deep-freeze or cryogenic storage in liquid or vapor phase liquid nitrogen, laboratory freezers, as well as transportation on dry ice. Resistant to multiple freeze-thaw cycles without fading, detaching, or wrinkling, these laser labels are provided in sheet format for printing with desktop and office laser printers, are wear-and-tear proof, will not jam printers, and resist UV and gamma irradiation.
Catalog No. NC1601260
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.