Learn More
Diagnostic Biosystems PERMABLACK/HRP PermaBlack/HRP
Supplier:  Diagnostic Biosystems K062
Encompass_Preferred
PermaBlack/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase. PermaBlack/HRP can be permanently mounted to produce a sharply contrasting ebony black color that can be easily distinguished from other stains.
Catalog No. 50-240-7624