Learn More
Diagnostic Biosystems PERMABLUE/HRP PermaBlue/HRP
Catalog No. 502407632
Encompass_Preferred
PermaBlue/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase. PermaBlue/HRP can be permanently mounted to produce a strong azure blue color that can be easily distinguished from other stains.
Catalog No. 50-240-7632 Supplier Diagnostic Biosystems Supplier No. K063110
PermaBlue/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase. PermaBlue/HRP can be permanently mounted to produce a strong azure blue color that can be easily distinguished from other stains.