Learn More
Diagnostic Biosystems PERMABLUE PLUS/AP PermaBlue Plus/AP
Catalog No. 502407633
Encompass_Preferred
PermaBlue Plus/AP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing alkaline phosphatase. PermaBlue Plus/AP has been reformulated to increase stability of the working solution while producing a distinct bright blue color. PermaBlue Plus/AP is insoluble in alcohol and xylene substitutes; therefore sections can be dehydrated in alcohol, cleared in xylene substitute, and permanently mounted.
Catalog No. 50-240-7633 Supplier Diagnostic Biosystems Supplier No. K058110
PermaBlue Plus/AP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing alkaline phosphatase. PermaBlue Plus/AP has been reformulated to increase stability of the working solution while producing a distinct bright blue color. PermaBlue Plus/AP is insoluble in alcohol and xylene substitutes; therefore sections can be dehydrated in alcohol, cleared in xylene substitute, and permanently mounted.