Learn More
Leica Biosystems PERMAFLEX Plus Adhesive Microscope Slides, Pink (MPN 3800019) - 75.5 x 25.5 x 1 mm, Case 1,440
Catalog No. NC2892237
Encompass_Preferred
Leica Biosystems PERMAFLEX Plus adhesive microscope slides (MPN 3800019) are pink, APES-coated slides engineered for reliable tissue adhesion and consistent imaging during automated staining, coverslipping, and digital scanning. Standard-size slides include a frosted labeling area for clear identification. Supplied in bulk packaging for high-volume laboratories.
  • Hydrophobic APES adhesive for improved tissue attachment.
  • Pink color for slide identification.
  • Standard dimensions: 75.50 ± 0.50 mm length x 25.50 ± 0.50 mm width x 1.00 ± 0.05 mm thickness.
  • Frosted labeling area: 18.50 ± 0.50 mm.
  • Bulk packaging: case 1,440 slides (box ~72 slides).
Catalog No. NC2892237 Supplier Leica Biosystems Supplier No. 3800019
Leica Biosystems PERMAFLEX Plus adhesive microscope slides (MPN 3800019) are pink, APES-coated slides engineered for reliable tissue adhesion and consistent imaging during automated staining, coverslipping, and digital scanning. Standard-size slides include a frosted labeling area for clear identification. Supplied in bulk packaging for high-volume laboratories.
  • Hydrophobic APES adhesive for improved tissue attachment.
  • Pink color for slide identification.
  • Standard dimensions: 75.50 ± 0.50 mm length x 25.50 ± 0.50 mm width x 1.00 ± 0.05 mm thickness.
  • Frosted labeling area: 18.50 ± 0.50 mm.
  • Bulk packaging: case 1,440 slides (box ~72 slides).