Learn More
Leica Biosystems PERMAFLEX PLUS ADHESIVE SLID
Catalog No. NC1647996
Encompass_Preferred
Newly designed frosted area (patent pending) is validated to work on the Leica IP S ink-jet and Perma S thermal printers.* Leica Biosystems PERMAFLEX and PERMAFLEX Plus adhesive slides are flexible print slides that show high print quality and readability by scanners, chemical resistance, positive staining and minimal background in five popular colors.PERMAFLEX BenefitsValidated to work on ink-jet and thermal printers**Tested for chemical resistance to common lab reagents (e.g. Xylene, chromic acid, etc.)Tested to provide consistent high-quality staining without any disadvantages of background stainingNewly designed color-coded packagingStandard sizing fits most instrumentation*Internally validated to work in the Leica IP S & Perma S as well as the ThermoFisher Slidemate printer.**Performance validation consisted of testing a total of 3,699 of all five colored slides on ink-jet and thermal printers.
This item is not returnable. View return policy
Catalog No. NC1647996 Supplier Leica Biosystems Supplier No. 3800014
Newly designed frosted area (patent pending) is validated to work on the Leica IP S ink-jet and Perma S thermal printers.* Leica Biosystems PERMAFLEX and PERMAFLEX Plus adhesive slides are flexible print slides that show high print quality and readability by scanners, chemical resistance, positive staining and minimal background in five popular colors.PERMAFLEX BenefitsValidated to work on ink-jet and thermal printers**Tested for chemical resistance to common lab reagents (e.g. Xylene, chromic acid, etc.)Tested to provide consistent high-quality staining without any disadvantages of background stainingNewly designed color-coded packagingStandard sizing fits most instrumentation*Internally validated to work in the Leica IP S & Perma S as well as the ThermoFisher Slidemate printer.**Performance validation consisted of testing a total of 3,699 of all five colored slides on ink-jet and thermal printers.