Learn More
Bio Plas PermaFrame 384-Well Full-Skirt PCR Plate with A24 Notch and Clear Wells
Catalog No. 22038933
Change view
Click to view available options
Color:
Assorted
Blue
Clear
Green
Red
Violet
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-933 Assorted
22-038-927 Clear
22-038-928 Blue
22-038-931 Violet
22-038-930 Red
22-038-932 Yellow
22-038-929 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-038-933 Supplier Bio Plas Supplier No. 9109
Add to Cart
Add to Cart
384-well PCR plate with full skirt.
  • PermaFrame 384-well
  • Full-skirt PCR plate
  • A24
  • Clear wells
Specifications
Barcode No Barcode
Format Standard
Array 24 x 16
Color Assorted
Disposable Single-use
Product Type PCR Plate
Material Polycarbonate
Volume (Metric) Well 30 μL
No. of Wells 384
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
RNase Free Yes
Fit for Purpose PCR Ready