Learn More
Bio Plas PermaFrame 96-Well Semi-Skirt PCR Plate with Clear Wells
Catalog No. 22038945
Change view
Click to view available options
Color:
Assorted
Blue
Clear
Green
Red
Violet
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-945 Violet
22-038-946 Yellow
22-038-941 Clear
22-038-942 Blue
22-038-943 Green
22-038-944 Red
22-038-947 Assorted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-038-945 Supplier Bio Plas Supplier No. 9305
Only null left
Add to Cart
Add to Cart
96-well PCR plate with semi-skirt.
  • PermaFrame 96-well semi-skirt
  • 0.2 mL PCR plate
  • Clear wells
Specifications
Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Not Autoclavable
Color Violet
Disposable Single-use
Product Type PCR Plate
Material Polycarbonate
Volume (Metric) Well 0.2 mL
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
RNase Free Yes
Fit for Purpose PCR Ready