Learn More
Diagnostic Biosystems PERMAGREEN/HRP PermaGreen/HRP
Catalog No. 502407443
Encompass_Preferred
PermaGreen™/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase. PermaGreen™/HRP can be permanently mounted to produce a bright green color that can be easily distinguished from other stains.
Catalog No. 50-240-7443 Supplier Diagnostic Biosystems Supplier No. K074110
PermaGreen™/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase. PermaGreen™/HRP can be permanently mounted to produce a bright green color that can be easily distinguished from other stains.