Learn More
Diagnostic Biosystems PERMAGREEN/HRP 30 ML PermaGreen/HRP
Catalog No. NC1742973
Encompass_Preferred
PermaGreen™/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase. PermaGreen™/HRP can be permanently mounted to produce a bright green color that can be easily distinguished from other stains.
Catalog No. NC1742973 Supplier Diagnostic Biosystems Supplier No. K074
PermaGreen™/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase. PermaGreen™/HRP can be permanently mounted to produce a bright green color that can be easily distinguished from other stains.