Learn More
Diagnostic Biosystems PERMAGREEN PLUS/AP PermaGreen Plus/AP
Catalog No. 502407634
Encompass_Preferred
PermaGreen Plus/AP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing alkaline phosphatase. PermaGreen Plus/AP has been modified to increase stability and staining intensity, producing astrong green color that is insoluble in alcohol and xylene substitutes (both aliphatic hydrocarbon and citrus based); therefore sections can be dehydrated in alcohol, cleared in xylene substitute*, and permanently mounted. However, we recommend air drying slides and then permanently mounting. This chromogen substrate system may be used for both automation and manual use.
Catalog No. 50-240-7634 Supplier Diagnostic Biosystems Supplier No. K059110
PermaGreen Plus/AP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing alkaline phosphatase. PermaGreen Plus/AP has been modified to increase stability and staining intensity, producing astrong green color that is insoluble in alcohol and xylene substitutes (both aliphatic hydrocarbon and citrus based); therefore sections can be dehydrated in alcohol, cleared in xylene substitute*, and permanently mounted. However, we recommend air drying slides and then permanently mounting. This chromogen substrate system may be used for both automation and manual use.