Learn More
GA International Permanent Autoclave-Resistant Labels for Laser Printers 2.63" x 1" / 66.8mm x 25.4mm Rectangle White From -40°C to +150°C (-40°F to +302°F)
Catalog No. 5000572010
Encompass_Preferred
Permanent laser printable labels, ideal for glassware, surgical instruments, metal and plastic surfaces, trays, blister packs, and other items intended for use in steam autoclaves, ovens, and gamma irradiators. Our autoclave laser labels are durable, inert, weather & water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121°C/250°F, 16.5 psi for 20 minutes). Compatible with all major brands of desktop laser printers.
Catalog No. 50-005-72010 Supplier GA International Supplier No. AKA3WH
Permanent laser printable labels, ideal for glassware, surgical instruments, metal and plastic surfaces, trays, blister packs, and other items intended for use in steam autoclaves, ovens, and gamma irradiators. Our autoclave laser labels are durable, inert, weather & water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121°C/250°F, 16.5 psi for 20 minutes). Compatible with all major brands of desktop laser printers.