Learn More
GA International Permanent Deep-Freeze Labels for Thermal-Transfer Barcode Printers 0.5" x 1.375" / 12.7mm x 34.9mm Rectangle White From -80°C to +110°C (-112°F to +230°F)
Catalog No. 5000572084
Encompass_Preferred
Deep-freeze, permanent barcode labels for small diameter vials, microtubes, falcon tubes, cell culture plates, microplates, as well as polypropylene and cardboard freezer boxes. The freezer-grade labels are waterproof and tearproof, can adhere to moist and wet surfaces, and will permanently adhere to containers in (-20°C, -40°C, -80°C) freezers and dry ice without the need for extra lamination. Available in multiple configurations and colors, they are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, surface cleaning wipes, and other chemicals.
Catalog No. 50-005-72084 Supplier GA International Supplier No. FJT104NPC11WH
Deep-freeze, permanent barcode labels for small diameter vials, microtubes, falcon tubes, cell culture plates, microplates, as well as polypropylene and cardboard freezer boxes. The freezer-grade labels are waterproof and tearproof, can adhere to moist and wet surfaces, and will permanently adhere to containers in (-20°C, -40°C, -80°C) freezers and dry ice without the need for extra lamination. Available in multiple configurations and colors, they are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, surface cleaning wipes, and other chemicals.