Learn More
GA International Permanent Deep-Freeze Labels for Thermal-Transfer Barcode Printers 0.94" x 0.5" + 0.437" (circle) / 23.9mm x 12.7mm + 11.1mm (circle) Rectangle + Circle Blue From -80°C to +110°C (-112°F to +230°F)
Catalog No. 5000571321
Encompass_Preferred
Deep-freeze, permanent barcode labels for small diameter vials, microtubes, falcon tubes, cell culture plates, microplates, as well as polypropylene and cardboard freezer boxes. The freezer-grade labels are waterproof and tearproof, can adhere to moist and wet surfaces, and will permanently adhere to containers in (-20°C, -40°C, -80°C) freezers and dry ice without the need for extra lamination. Available in multiple configurations and colors, they are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, surface cleaning wipes, and other chemicals.
Catalog No. 50-005-71321 Supplier GA International Supplier No. FJT52NOTC12BL
Deep-freeze, permanent barcode labels for small diameter vials, microtubes, falcon tubes, cell culture plates, microplates, as well as polypropylene and cardboard freezer boxes. The freezer-grade labels are waterproof and tearproof, can adhere to moist and wet surfaces, and will permanently adhere to containers in (-20°C, -40°C, -80°C) freezers and dry ice without the need for extra lamination. Available in multiple configurations and colors, they are made of a durable thermoplastic film and will resist spraying with alcohols, bleach, detergents, surface cleaning wipes, and other chemicals.