Learn More
Cole-Parmer Permanent Fine-Point Paint Markers, Black; 12/Box
Supplier:  Cole-Parmer 0996461
Encompass
Write on almost any surface—metal, glass, plastic, cardboard, or wood. Ideal for color coding with seven colors available; use white or yellow for dark surfaces. Unique valve prevents tip from drying out.
Catalog No. NC9015793