Learn More
Electron Microscopy Sciences Permanox Dish, 60 x 15 mm
SDP
Supplier:  Electron Microscopy Sciences 70353
Encompass_Preferred
Permanox is a strong, biologically inert material, resistant to nonchlorinated hydrocarbons, acetone, and many other EM reagents. Our EMbed-812 and other embedding resins can be used to embed cells in these dishes with excellent results.
Permanox has not been shown to be autofluorescent, and has a low oxygen solubility which makes it suitable for experiments performed under anoxic conditions.
The dishes have high optical quality that provide excellent and consistent viewing where microscopic examination is required.
Size: 60 x 15 mm
Catalog No. 50-930-209