Learn More
Electron Microscopy Sciences Permanox Dish, 60 x 15 mm
SDP
Catalog No. 501899245
Encompass_Preferred
Permanox is a strong, biologically inert material, resistant to nonchlorinated hydrocarbons, acetone, and many other EM reagents. Our EMbed-812 and other embedding resins can be used to embed cells in these dishes with excellent results.
Permanox has not been shown to be autofluorescent, and has a low oxygen solubility which makes it suitable for experiments performed under anoxic conditions.
The dishes have high optical quality that provide excellent and consistent viewing where microscopic examination is required.
Size: 60 x 15 mm
Catalog No. 50-189-9245 Supplier Electron Microscopy Sciences Supplier No. 70356
Permanox is a strong, biologically inert material, resistant to nonchlorinated hydrocarbons, acetone, and many other EM reagents. Our EMbed-812 and other embedding resins can be used to embed cells in these dishes with excellent results.
Permanox has not been shown to be autofluorescent, and has a low oxygen solubility which makes it suitable for experiments performed under anoxic conditions.
The dishes have high optical quality that provide excellent and consistent viewing where microscopic examination is required.
Size: 60 x 15 mm