Learn More
Electron Microscopy Sciences Microscope Slide Microscope Slide Permanox
SDP
Catalog No. 50949438
Encompass_Preferred
Permanox is a strong, biologically inert material resistant to non-chlorinated hydrocarbons, acetone, and other EM reagents. Permanox slides allow you to do cell fixation and staining on the slide and they can be cut with scissors. Permanox has not been shown to be autofluorescent. All of the microscope slides are 1" x 3".
NOTE: Long exposure to solvents will cause the slide to warp. Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.
Catalog No. 50-949-438 Supplier Electron Microscopy Sciences Supplier No. 71885
Permanox is a strong, biologically inert material resistant to non-chlorinated hydrocarbons, acetone, and other EM reagents. Permanox slides allow you to do cell fixation and staining on the slide and they can be cut with scissors. Permanox has not been shown to be autofluorescent. All of the microscope slides are 1" x 3".
NOTE: Long exposure to solvents will cause the slide to warp. Xylene and Toluene based mounting media should be avoided. Glycerol, gelatin, and other aqueous based media are recommended.