Learn More
Diagnostic Biosystems PERMARED/AP PermaRed/AP
Catalog No. 502407531
Encompass_Preferred
PermaRed/AP is a substrate-chromogen system designed to be used for either IHC or ISH when using alkaline phosphatase detection. PermaRed/AP produces a brilliant dark red color. PermaRed/AP is insoluble in organic solvents; therefore, sections can be dehydrated in alcohol, cleared in xylene (or a xylene-substitute), and permanently mounted. This chromogen substrate system may be used for both automation and manual use.
Catalog No. 50-240-7531 Supplier Diagnostic Biosystems Supplier No. K049110
PermaRed/AP is a substrate-chromogen system designed to be used for either IHC or ISH when using alkaline phosphatase detection. PermaRed/AP produces a brilliant dark red color. PermaRed/AP is insoluble in organic solvents; therefore, sections can be dehydrated in alcohol, cleared in xylene (or a xylene-substitute), and permanently mounted. This chromogen substrate system may be used for both automation and manual use.