Learn More
Diagnostic Biosystems PERMARED/AP ALCOHOLXYLENE PermaRed/AP
Catalog No. NC2049152
Encompass_Preferred
PermaRed/AP is a substrate-chromogen system designed to be used for either IHC or ISH when using alkaline phosphatase detection. PermaRed/AP produces a brilliant dark red color. PermaRed/AP is insoluble in organic solvents; therefore, sections can be dehydrated in alcohol, cleared in xylene (or a xylene-substitute), and permanently mounted. This chromogen substrate system may be used for both automation and manual use.
This item is not returnable. View return policy
Catalog No. NC2049152 Supplier Diagnostic Biosystems Supplier No. K049
PermaRed/AP is a substrate-chromogen system designed to be used for either IHC or ISH when using alkaline phosphatase detection. PermaRed/AP produces a brilliant dark red color. PermaRed/AP is insoluble in organic solvents; therefore, sections can be dehydrated in alcohol, cleared in xylene (or a xylene-substitute), and permanently mounted. This chromogen substrate system may be used for both automation and manual use.