Learn More
Newcomer Supply Permaslip Mounting Medium & Liquid Coverslip, 8 oz
SDP
Supplier:  Newcomer Supply 6530B
Encompass
Newcomer Supply Permaslip Mounting Medium is reliable and easy-to-use with excellent results as a standard mounting medium product or as a liquid coverslip (coverglass free) mounting medium. It is a clear acrylic medium that is fast drying. Permaslip Mounting Medium can also be used in tissue/cell transfer procedures to transfer tissue sections or cell preparations from a prepared microscopic slide when specimen availability is limited and additional diagnostic workup is necessary. Examples when tissue/cell transfer with Permaslip is applicable include: Paraffin sections of small biopsies/minimal tissue, Biopsies with a small foci of neoplasm, Paraffin block is unavailable, Cytology preparations, fine needle aspirates (FNA), Transfer of tissue from an uncharged slide to charged slide(s), Preparing multiple slides from a single slide preparation, Salvaging/restoring broken slides back to a whole mount. Unique spout on 8 oz cans controls drop by drop application.
Catalog No. NC0530667