Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wearwell™ PermaTack™ Contamination Control Mats

Catalog No. 19151144 Shop All Wearwell Tennessee Mat Company Inc. Products
Change view
Click to view available options
Dimensions (L x W):
36 x 54 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-151-144 36 x 54 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-151-144 Supplier Wearwell™ Supplier No. 092.18X36X54BL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

A high-performance, industrial strength contamination control mat

  • Permanently tacky surface removes even the finest particles off shoes or wheels
  • 98% to 100% of contamination will be removed if each foot touches the mat three times
  • Anti-microbial blue vinyl polymer
  • High performance backing provides structural durability
  • Unique adhesive backing bonds to the floor

Warranty and Services

One year.

Specifications

Color Blue
Dimensions (L x W) 36 x 54 in.
For Use With (Application) Removing particles off shoes or wheels
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.