Learn More
Diagnostic Biosystems PERMAYELLOW/HRP PermaYellow/HRP
Catalog No. 502407532
Encompass_Preferred
PermaYellow/HRP is a substrate chromogen system for use with horseradish peroxidase (HRP) detection in immunohistochemistry (IHC) or in situ hybridizations (ISH). PermaYellow/HRP produces a distinct bright yellow that can easily be distinguished from other stains.
Catalog No. 50-240-7532 Supplier Diagnostic Biosystems Supplier No. K060110
PermaYellow/HRP is a substrate chromogen system for use with horseradish peroxidase (HRP) detection in immunohistochemistry (IHC) or in situ hybridizations (ISH). PermaYellow/HRP produces a distinct bright yellow that can easily be distinguished from other stains.