Learn More
Alkali Scientific ASI™ Permea™ Dish, Easy Grip, Sterile, 50/Cs
SDP
Catalog No. 503027901
Encompass_Preferred
The ASIPermea™ Dish ensures optimal gas exchange for effective cell culture. Due to its low water permeability, the culture medium is maintained for a long time without leakage or evaporation.  The ASIPermea™ Dish has low auto-fluorescence with high light transmission to enable applications in imaging and micro-reading   • Ergonomic design for easy handling and minimal contamination  • Gas-permeability tested (CO2, O2) • Leakage and evaporation tested • Transparent for microscopic observation and imaging • Low auto-fluorescence • High light transmission • Sterile ▪ Non-cytotoxic▪ Non-pyrogenic▪ DNase / RNase – free▪ Human DNA – free
Catalog No. 50-302-7901 Supplier Alkali Scientific Supplier No. SPD4488
The ASIPermea™ Dish ensures optimal gas exchange for effective cell culture. Due to its low water permeability, the culture medium is maintained for a long time without leakage or evaporation.  The ASIPermea™ Dish has low auto-fluorescence with high light transmission to enable applications in imaging and micro-reading   • Ergonomic design for easy handling and minimal contamination  • Gas-permeability tested (CO2, O2) • Leakage and evaporation tested • Transparent for microscopic observation and imaging • Low auto-fluorescence • High light transmission • Sterile ▪ Non-cytotoxic▪ Non-pyrogenic▪ DNase / RNase – free▪ Human DNA – free