Learn More
Enzo Life Sciences Permeabilization, (buffer) (100 ml)
SDP
Catalog No. 50658673
Encompass_Preferred
Formulation: Liquid. PBS, pH 7.2, containing 0.1% saponin and 0.09% sodium azide. Applications: Flow Cytometry. Shipping: Blue Ice Not Frozen. Long Term Storage: +4°C
Catalog No. 50-658-673 Supplier Enzo Life Sciences Supplier No. ADI950012100
Formulation: Liquid. PBS, pH 7.2, containing 0.1% saponin and 0.09% sodium azide. Applications: Flow Cytometry. Shipping: Blue Ice Not Frozen. Long Term Storage: +4°C