Learn More
Abcam Permeabilization Buffer (Flow Cytometry), 100ML
Catalog No. 502699021
Encompass_Preferred
Permeabilization Buffer (ab314681) is a ready to use permeabilization buffer for intracellular immunofluorescence staining for microscopy or flow cytometry.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-9021 Supplier Abcam Supplier No. AB314681100ML
Permeabilization Buffer (ab314681) is a ready to use permeabilization buffer for intracellular immunofluorescence staining for microscopy or flow cytometry.
The product is subject to the following: Abcam Restricted Use Statement