Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Permeabilization Medium (Medium B)

Catalog No. GAS002S100
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GAS002S100 100 mL
GAS002S5 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. GAS002S100 Supplier Invitrogen™ Supplier No. GAS002S100
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Permeabilization Medium (Medium B)

Permeabilization Medium (Medium B) is a component of the FIX & PERM™ Cell Permeabilization Kit. The kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics.

The FIX & PERM™ Cell Permeabilization Kit is:

  • Flexible—compatible with multiple sample types and cellular antigens
  • Robust—scatter pattern of cells unaffected
  • Efficient—simultaneous detection of intracellular and surface markers

    View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).

    The Permeabilization Medium formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.

    How It Works
    Cells are incubated for 15 minutes at room temperature with Fixation Medium (Medium A) (Cat. No. GAS001S100) and washed. Cells are then incubated with the Permeabilization Medium and fluorochrome-labeled antibody. Both Fixation Medium and Permeabilization Medium are included in the FIX & PERM™ Cell Permeabilization Kit.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Room Temperature
    Type Permeabilization Medium (Medium B)
    Product Line FIX & PERM
    Quantity 100 mL
    Reagent Type Permeabilization Reagent
    Shipping Condition Room Temperature
    The FIX & PERM Cell Permeabilization Kit protocol does not say what volume my cells should be in when they are added to the tube. What should I do?

    Your cells should be in approximately 100 µL volume. One way to do this is to aliquot the correct number of cells (10E6) and then spin them down and decant the supernatant. The residual volume should be approximately 100 µL.

    Can I use the FIX & PERM Cell Permeabilization Kit to access nuclear epitopes?

    Yes. Generally, you need to increase the incubation time in the permeabilization buffer (Reagent B). Here are a couple of references:
    -Millard I, Degrave E, Philippe M et al. (1998) Detection of intracellular antigens by flow cytometry: comparison of two chemical methods and microwave heating. Clin Chem 44:2320-2330(http://www.ncbi.nlm.nih.gov/pubmed/9799760).
    -Parker JE, Mufti GJ, Rasool F et al. (2000) The role of apoptosis, proliferation, and the Bcl-2-related proteins in the myelodysplastic syndromes and acute myeloid leukemia secondary to MDS. Blood 96:3932-3938(http://www.ncbi.nlm.nih.gov/pubmed/11090080).

    The FIX & PERM Cell Permeabilization Kit is used to prepare cells for flow cytometry; can I use it for immunohistochemistry?

    Yes. Here are a couple of references:
    Haranaga S, Yamaguchi H, Friedman H et al.(2001) Chlamydia pneumoniae infects and multiplies in lymphocytes In vitro. Infect Immun 69:7753-7759 (http://www.ncbi.nlm.nih.gov/pubmed/11705957).

    Al-Mufti R, Hambley H, Albaiges G et al. (2000) Increased fetal erythroblasts in women who subsequently develop pre-eclampsia. Hum Reprod 15:1624-1628 (http://www.ncbi.nlm.nih.gov/pubmed/10875878).

    Does the FIX & PERM Cell Permeabilization Kit (Cat. Nos. GAS003 and GAS004) lyse red blood cells?

    Yes. The FIX & PERM Cell Permeabilization Kit is used in flow cytometry for fixing and permeabilizing cells in suspension, allowing access of antibody to intracellular targets without sacrificing scatter characteristics. It has been validated for use in cell lines, whole blood, and bone marrow. In addition fixing and permeabilizing cells, it lyses red blood cells. It is labeled as a general purpose reagent (GPR) and can be used in clinical settings, but it is also used widely in research labs.

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.