Learn More
Bellco Glass, Inc. Permeable Cell Culture Inserts
SDP
Catalog No. 502329246
Encompass_Preferred
Permeable Cell Culture Inserts mimic in vivo conditions. Inserts center in wells preventing membrane from touching sides and wicking between insert and side wall. Membrane remains suspended above well bottom allowing coculture grow one cell line inside insert and another below in the well. Ideal for drug transfer assays transport studies migration assays and cellcell interaction studies. NonTreated Plates sold separately carrier plate is Tissue Culture Treated which encourages cell adhesion.Specifications 0.4µm pore size is ideal for coculture and drug transport studies 3.0µm and 8.0µm pore size are ideal for angiogenesis and chemotaxis experiments Membrane Material PC Membrane Pore Size 0.4um Case Quantity 12
Catalog No. 50-232-9246 Supplier Bellco Glass, Inc. Supplier No. 230601000
Permeable Cell Culture Inserts mimic in vivo conditions. Inserts center in wells preventing membrane from touching sides and wicking between insert and side wall. Membrane remains suspended above well bottom allowing coculture grow one cell line inside insert and another below in the well. Ideal for drug transfer assays transport studies migration assays and cellcell interaction studies. NonTreated Plates sold separately carrier plate is Tissue Culture Treated which encourages cell adhesion.Specifications 0.4µm pore size is ideal for coculture and drug transport studies 3.0µm and 8.0µm pore size are ideal for angiogenesis and chemotaxis experiments Membrane Material PC Membrane Pore Size 0.4um Case Quantity 12