Learn More
Logan Instruments Corp PermeaPlain Premium Plate, 96 Wells, Uncoated Regenerated Cellulose, 5 Plates
Catalog No. NC3345706
Encompass
The Logan Instruments Corp. PermeaPlain Premium Plate (5/PK) is a high-throughput multi-well plate system designed for passive mass transfer analysis in drug research. It features a unique uncoated regenerated cellulose hydrate membrane that allows for efficient drug diffusion studies, regardless of polarity, providing reproducible results for key parameters such as permeation and flux. This product is suitable for a variety of analysis methods, making it an essential tool for enhancing drug development processes.
Catalog No. NC3345706 Supplier Logan Instruments Corp Supplier No. 07800085
The Logan Instruments Corp. PermeaPlain Premium Plate (5/PK) is a high-throughput multi-well plate system designed for passive mass transfer analysis in drug research. It features a unique uncoated regenerated cellulose hydrate membrane that allows for efficient drug diffusion studies, regardless of polarity, providing reproducible results for key parameters such as permeation and flux. This product is suitable for a variety of analysis methods, making it an essential tool for enhancing drug development processes.