Learn More
Chem Service Inc Permethrin,52645-53-1,250MG
SDP
Catalog No. NC9865654
Encompass_Preferred
Molecular Formula C21H20Cl2O3; Purity 99.5%; Molecular Weight 391.29; Boiling Point > 290 °C
Catalog No. NC9865654 Supplier Chem Service Inc Supplier No. N12848250MG
Molecular Formula C21H20Cl2O3; Purity 99.5%; Molecular Weight 391.29; Boiling Point > 290 °C