Learn More
Enzo Life Sciences Permethrin (50mg). CAS: 52645-53-1
SDP
Catalog No. 502011501
Encompass_Preferred
Synthetic type I pyrethroid. Structurally similar to the protein phosphatase 2B (calcineurin) inhibitors cypermethrin and deltamethrin but weakly active or inactive depending on assay conditions. May be used as a negative control for cypermethrin (BML-PR100) and deltamethrin (BML-PR101). Alternative name: 3-Phenoxybenzyl (1R,S)-cis,trans-3-(2,2-dichlorovinyl)-2,2-dimethylcyclo-propanecarboxylate. Purity: ≥96% (HPLC) . Solubility: Soluble in DMSO (40mg/ml) or ethanol (25mg/ml). Insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-1501 Supplier Enzo Life Sciences Supplier No. BMLPR1030050
Synthetic type I pyrethroid. Structurally similar to the protein phosphatase 2B (calcineurin) inhibitors cypermethrin and deltamethrin but weakly active or inactive depending on assay conditions. May be used as a negative control for cypermethrin (BML-PR100) and deltamethrin (BML-PR101). Alternative name: 3-Phenoxybenzyl (1R,S)-cis,trans-3-(2,2-dichlorovinyl)-2,2-dimethylcyclo-propanecarboxylate. Purity: ≥96% (HPLC) . Solubility: Soluble in DMSO (40mg/ml) or ethanol (25mg/ml). Insoluble in water. Long Term Storage: Ambient.