Learn More
Medchemexpress LLC Permethrin-d5 | 1794760-19-2 | 99.2% | 1 MG
SDP
Catalog No. 5000408709
Encompass_Preferred
Permethrin-d5 (NRDC-143-d5) is the deuterium labeled Permethrin. Permethrin (NRDC-143) is an insecticide, acaricide, and insect repellent; it functions as a neurotoxin by affecting neuron membranes and prolonging sodium channel activation. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08709 Supplier Medchemexpress LLC Supplier No. HYB0887S1MG
Permethrin-d5 (NRDC-143-d5) is the deuterium labeled Permethrin. Permethrin (NRDC-143) is an insecticide, acaricide, and insect repellent; it functions as a neurotoxin by affecting neuron membranes and prolonging sodium channel activation. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.