Learn More
Medchemexpress LLC Permethrin-d9 | 52645-53-1 | 98.2% | 1 MG
SDP
Catalog No. 5000408666
Encompass_Preferred
Permethrin-d9 is a deuterium-labeled version of Permethrin, an insecticide, acaricide, and insect repellent. It functions as a neurotoxin by affecting neuron membranes through prolonging sodium channel activation. This compound is for research use only and can be utilized as a tracer or an internal standard for quantitative analysis.
  • Deuterium-labeled Permethrin
  • Functions as an insecticide, acaricide, and insect repellent
  • Acts as a neurotoxin by prolonging sodium channel activation
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuteration can influence pharmacokinetic and metabolic profiles
Catalog No. 50-004-08666 Supplier Medchemexpress LLC Supplier No. HYB0887S11MG
Permethrin-d9 is a deuterium-labeled version of Permethrin, an insecticide, acaricide, and insect repellent. It functions as a neurotoxin by affecting neuron membranes through prolonging sodium channel activation. This compound is for research use only and can be utilized as a tracer or an internal standard for quantitative analysis.
  • Deuterium-labeled Permethrin
  • Functions as an insecticide, acaricide, and insect repellent
  • Acts as a neurotoxin by prolonging sodium channel activation
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuteration can influence pharmacokinetic and metabolic profiles