Learn More
Medchemexpress LLC Permethrin-d9 | 52645-53-1 | 98.2% | 5 MG
SDP
Catalog No. 5000409269
Encompass_Preferred
Permethrin-d9 is a deuterium labeled Permethrin. Permethrin functions as a neurotoxin, affecting neuron membranes by prolonging sodium channel activation, and is used as an insecticide, acaricide, and insect repellent.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Suitable as an isotope-labeled compound.
  • Useful as an internal standard for food, pesticides, and veterinary medicine.
  • Applicable as an internal standard for environmental contaminants.
Catalog No. 50-004-09269 Supplier Medchemexpress LLC Supplier No. HYB0887S15MG
Permethrin-d9 is a deuterium labeled Permethrin. Permethrin functions as a neurotoxin, affecting neuron membranes by prolonging sodium channel activation, and is used as an insecticide, acaricide, and insect repellent.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Suitable as an isotope-labeled compound.
  • Useful as an internal standard for food, pesticides, and veterinary medicine.
  • Applicable as an internal standard for environmental contaminants.