Learn More
Medchemexpress LLC Perospirone hydrochloride | 129273-38-7 | MFCD00890882 | 99.9% | 463.04 g/mol | C23H31ClN4O2S | 10 MG
SDP
Catalog No. 5000297376
Encompass_Preferred
Perospirone hydrochloride is the hydrochloride salt of perospirone (SM-9018), an atypical antipsychotic research compound that antagonizes 5-HT2A and dopamine D2 receptors and acts as a partial agonist at 5-HT1A. It is supplied for preclinical pharmacology and biochemical research into serotonergic and dopaminergic signaling.
  • High chemical purity suitable for research (reported 99.94%).
  • Hydrochloride salt form for improved stability and handling.
  • Molecular weight 463.04 g/mol; molecular formula C23H31ClN4O2S.
  • Used in receptor binding and schizophrenia-related pharmacology studies.
  • Available in small milligram pack sizes for laboratory use.
Catalog No. 50-002-97376 Supplier Medchemexpress LLC Supplier No. HYB073110MG
Perospirone hydrochloride is the hydrochloride salt of perospirone (SM-9018), an atypical antipsychotic research compound that antagonizes 5-HT2A and dopamine D2 receptors and acts as a partial agonist at 5-HT1A. It is supplied for preclinical pharmacology and biochemical research into serotonergic and dopaminergic signaling.
  • High chemical purity suitable for research (reported 99.94%).
  • Hydrochloride salt form for improved stability and handling.
  • Molecular weight 463.04 g/mol; molecular formula C23H31ClN4O2S.
  • Used in receptor binding and schizophrenia-related pharmacology studies.
  • Available in small milligram pack sizes for laboratory use.