Learn More
Sigma Aldrich Perovskite quantum dots
Catalog No. 9007485ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
90-074-85ML 5 mL
1 options
Catalog No. 90-074-85ML Supplier Sigma Aldrich Supplier No. 9007485ML
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
For Perovskite quantum dots:Perovskite quantum dots (QDs) of common formula CsPbX3 (X = Cl, Br, I) possess high photoluminescence efficiency and narrow emission and emit in the visible spectral regime. Perovskite QDs are cadmium free and the aforementioned properties render them suitable for applications in light emitting diodes (LEDs), lasers, liquid crystal displays (LCDs) etc.
Specifications
Quantity 5 mL
Synonym Cadmium free QDs; Fluorescent nanocrystals; Perovskite nanocrystals; QDs
Biological Activity Central nervous system