Learn More
Jackson Immuno Research Labs Peroxidase AffiniPure Goat Anti-Rat IgG, Fcγ fragment specific, 1.5ml
SDP
Supplier:  Jackson Immuno Research Labs 112035071
Encompass_Preferred
Peroxidase-AffiniPure Goat Anti-Rat IgG, Fc Fragment Specific (min X Hu,Bov,Hrs Sr Prot)
Catalog No. NC9733959