Learn More
Biocare Medical, LLC PeroxAbolish - 100 ml
SDP
Catalog No. 5083255
Encompass_Preferred
PeroxAbolish is a novel formula that is a non-hydrogen peroxide blocking reagent that is safe and extremely gentle on tissues and cells. It is highly effective in quenching endogenous peroxidase and can be used on formalin-fixed paraffin-embedded tissues, cell culture, blood smears, cell preparations and frozen sections. When using paraffin-embedded formalin-fixed tissues, PeroxAbolish is extremely effective after antigen retrieval methods. It can also be used on an automated staining system.
Catalog No. 50-832-55 Supplier Biocare Medical, LLC Supplier No. PXA969L
 Restricted Item
Add to Cart
Add to Cart
PeroxAbolish is a novel formula that is a non-hydrogen peroxide blocking reagent that is safe and extremely gentle on tissues and cells. It is highly effective in quenching endogenous peroxidase and can be used on formalin-fixed paraffin-embedded tissues, cell culture, blood smears, cell preparations and frozen sections. When using paraffin-embedded formalin-fixed tissues, PeroxAbolish is extremely effective after antigen retrieval methods. It can also be used on an automated staining system.