Learn More
Jackson Immuno Research Labs Peroxidase AffiniPure Goat Anti-Alpaca IgG, subclasses 2+3 specific, 0.25 mL
SDP
Catalog No. NC1681438
Encompass_Preferred
Based on immunoelectrophoresis and/or ELISA, the antibody reacts with heavy chain (HC) alpaca IgG, subclasses 2 and 3. It also reacts with HC llama IgG, subclasses 2 and 3. No antibody was detected against alpaca or llama IgG1, light chains or non-immunoglobulin serum proteins. The antibody reacts primarily with the Fc region, and is not recommended for detection of VHH antibodies. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, human, mouse, rabbit, and rat serum proteins, but it may cross-react with immunoglobulins from other species.
Catalog No. NC1681438 Supplier Jackson Immuno Research Labs Supplier No. 128035229
Based on immunoelectrophoresis and/or ELISA, the antibody reacts with heavy chain (HC) alpaca IgG, subclasses 2 and 3. It also reacts with HC llama IgG, subclasses 2 and 3. No antibody was detected against alpaca or llama IgG1, light chains or non-immunoglobulin serum proteins. The antibody reacts primarily with the Fc region, and is not recommended for detection of VHH antibodies. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, human, mouse, rabbit, and rat serum proteins, but it may cross-react with immunoglobulins from other species.